Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPM1D
Entrez Gene ID
101108932
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1D
Gene Aliases
-
Chromosome Location
Chr.11: 51566622 - 51626405 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027975327.1 XP_027831128.1 5 - 6 1419 87

View Full Product Details