Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SAMD15
Entrez Gene ID
101110598
Gene Name
sterile alpha motif domain containing 15
Gene Aliases
-
Chromosome Location
Chr.7: 92583394 - 92594000 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004011085.4 XP_004011134.1 1 - 2 1791 72

View Full Product Details