Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KRT36
Entrez Gene ID
101117175
Gene Name
keratin 36
Gene Aliases
-
Chromosome Location
Chr.11: 21435836 - 21439860 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004012900.4 XP_004012949.2 6 - 7 1262 119

View Full Product Details