Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NPTXR
Entrez Gene ID
105609482
Gene Name
neuronal pentraxin receptor
Gene Aliases
-
Chromosome Location
Chr.3: 231251388 - 231272915 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015095030.2 XP_014950516.2 2 - 3 854 78

View Full Product Details