Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BEND4
Entrez Gene ID
101111023
Gene Name
BEN domain containing 4
Gene Aliases
-
Chromosome Location
Chr.6: 66518620 - 66547570 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027971395.1 XP_027827196.1 3 - 4 1139 105

View Full Product Details