Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLF1
Entrez Gene ID
101108386
Gene Name
SMC5-SMC6 complex localization factor 1
Gene Aliases
ANKRD32
Chromosome Location
Chr.5: 99789413 - 99862620 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027970521.1 XP_027826322.1 2 - 3 200 96

View Full Product Details