Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRIML1
Entrez Gene ID
101117391
Gene Name
tripartite motif family like 1
Gene Aliases
-
Chromosome Location
Chr.26: 18648099 - 18654695 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004021722.4 XP_004021771.3 5 - 6 859 66

View Full Product Details