Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GOT1L1
Entrez Gene ID
101111158
Gene Name
glutamic-oxaloacetic transaminase 1 like 1
Gene Aliases
-
Chromosome Location
Chr.26: 36156364 - 36162294 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004021775.4 XP_004021824.1 3 - 4 952 88

View Full Product Details