Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RFXAP
Entrez Gene ID
105612445
Gene Name
regulatory factor X associated protein
Gene Aliases
-
Chromosome Location
Chr.10: 27276473 - 27285162 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012185000.3 XP_012040390.3 2 - 3 794 116

View Full Product Details