Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRPR
Entrez Gene ID
100142675
Gene Name
gastrin releasing peptide receptor
Gene Aliases
-
Chromosome Location
Chr.X: 15013963 - 15050340 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004021935.4 XP_004021984.1 1 - 2 410 97

View Full Product Details