Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHL36
Entrez Gene ID
101103833
Gene Name
kelch like family member 36
Gene Aliases
-
Chromosome Location
Chr.14: 11511441 - 11527420 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027977689.1 XP_027833490.1 3 - 4 1481 75
XM_027977690.1 XP_027833491.1 3 - 4 1572 75

View Full Product Details