Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD33B
Entrez Gene ID
101105441
Gene Name
ankyrin repeat domain 33B
Gene Aliases
-
Chromosome Location
Chr.16: 68561478 - 68662652 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027980167.1 XP_027835968.1 3 - 4 641 101

View Full Product Details