Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
FAM83D
Entrez Gene ID
101102582
Gene Name
family with sequence similarity 83 member D
Gene Aliases
-
Chromosome Location
Chr.13: 70532926 - 70553208 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015099882.2 XP_014955368.2 2 - 3 674 96

View Full Product Details