Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR72
Entrez Gene ID
101101895
Gene Name
WD repeat domain 72
Gene Aliases
-
Chromosome Location
Chr.7: 58368084 - 58563036 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027971818.1 XP_027827619.1 11 - 12 1574 74

View Full Product Details