Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD49
Entrez Gene ID
101115060
Gene Name
ankyrin repeat domain 49
Gene Aliases
-
Chromosome Location
Chr.15: 1240857 - 1259938 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004015956.4 XP_004016005.1 2 - 3 444 73
XM_012095277.3 XP_011950667.1 2 - 3 454 73
XM_012095278.3 XP_011950668.1 2 - 3 483 73

View Full Product Details