Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SRP9
Entrez Gene ID
101110607
Gene Name
signal recognition particle 9
Gene Aliases
-
Chromosome Location
Chr.12: 31493079 - 31503698 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004013626.4 XP_004013675.1 1 - 2 183 80

View Full Product Details