Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NDUFC1
Entrez Gene ID
101116510
Gene Name
NADH:ubiquinone oxidoreductase subunit C1
Gene Aliases
-
Chromosome Location
Chr.17: 19898340 - 19902965 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004017243.3 XP_004017292.1 2 - 3 211 96

Gene Symbol
LOC114112897
Entrez Gene ID
114112897
Gene Name
NADH dehydrogenase [ubiquinone] 1 subunit C1, mitochondrial
Gene Aliases
-
Chromosome Location
Chr.2: 239912339 - 239912744 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027963981.1 XP_027819782.1 1 - 1 170 96

View Full Product Details