Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NKG7
Entrez Gene ID
101115817
Gene Name
natural killer cell granule protein 7
Gene Aliases
-
Chromosome Location
Chr.14: 61170631 - 61172323 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027977537.1 XP_027833338.1 1 - 2 408 65

View Full Product Details