Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
TREM1
Entrez Gene ID
101105695
Gene Name
triggering receptor expressed on myeloid cells 1
Gene Aliases
-
Chromosome Location
Chr.20: 16891729 - 16906014 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027958398.1 XP_027814199.1 3 - 3 552 105
XM_027958399.1 XP_027814200.1 3 - 3 431 105

View Full Product Details