Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HAPLN4
Entrez Gene ID
101105570
Gene Name
hyaluronan and proteoglycan link protein 4
Gene Aliases
-
Chromosome Location
Chr.5: 4054401 - 4064393 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027969218.1 XP_027825019.1 4 - 5 720 90

View Full Product Details