Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSHB
Entrez Gene ID
101104284
Gene Name
thyroid stimulating hormone subunit beta
Gene Aliases
-
Chromosome Location
Chr.1: 97229509 - 97234751 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004002368.4 XP_004002417.2 1 - 2 41 86
GenBank mRNA X90775.1 - - 47 86

View Full Product Details