Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTX3
Entrez Gene ID
100034672
Gene Name
pentraxin 3
Gene Aliases
-
Chromosome Location
Chr.1: 250842659 - 250849671 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004003220.4 XP_004003269.2 1 - 2 1206 100

View Full Product Details