Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ICAM4
Entrez Gene ID
101116476
Gene Name
intercellular adhesion molecule 4 (Landsteiner-Wiener blood group)
Gene Aliases
-
Chromosome Location
Chr.5: 13157863 - 13159311 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004009292.3 XP_004009341.2 1 - 2 677 59

View Full Product Details