Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VSIG10L
Entrez Gene ID
101122129
Gene Name
V-set and immunoglobulin domain containing 10 like
Gene Aliases
-
Chromosome Location
Chr.14: 61136978 - 61151169 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027977539.1 XP_027833340.1 1 - 2 225 106

View Full Product Details