Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EMP1
Entrez Gene ID
101109432
Gene Name
epithelial membrane protein 1
Gene Aliases
-
Chromosome Location
Chr.3: 216906624 - 216928296 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004006842.4 XP_004006891.1 4 - 5 498 93

View Full Product Details