Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
B3GAT3
Entrez Gene ID
101111917
Gene Name
beta-1,3-glucuronyltransferase 3
Gene Aliases
-
Chromosome Location
Chr.21: 42822949 - 42827636 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004019603.4 XP_004019652.1 1 - 2 174 78

View Full Product Details