Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ESM1
Entrez Gene ID
101117876
Gene Name
endothelial cell specific molecule 1
Gene Aliases
-
Chromosome Location
Chr.16: 25486840 - 25494692 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004016994.4 XP_004017043.2 1 - 2 645 67

View Full Product Details