Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CSNK2B
Entrez Gene ID
100216435
Gene Name
casein kinase 2 beta
Gene Aliases
-
Chromosome Location
Chr.20: 29464215 - 29470511 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027958334.1 XP_027814135.1 1 - 2 1258 67

View Full Product Details