Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCLK3
Entrez Gene ID
101114472
Gene Name
doublecortin like kinase 3
Gene Aliases
-
Chromosome Location
Chr.19: 11324837 - 11367227 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027958150.1 XP_027813951.1 2 - 3 1909 80

View Full Product Details