Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPM1N
Entrez Gene ID
101117357
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1N (putative)
Gene Aliases
-
Chromosome Location
Chr.14: 56588660 - 56593060 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015100599.2 XP_014956085.2 4 - 5 1226 59

View Full Product Details