Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C15H11orf16
Entrez Gene ID
101106876
Gene Name
chromosome 15 C11orf16 homolog
Gene Aliases
-
Chromosome Location
Chr.15: 46810140 - 46819412 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004016173.4 XP_004016222.3 7 - 8 1679 74

View Full Product Details