Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MS4A5
Entrez Gene ID
101113009
Gene Name
membrane spanning 4-domains A5
Gene Aliases
-
Chromosome Location
Chr.15: 89547112 - 89574558 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004016512.3 XP_004016561.1 4 - 5 495 81

View Full Product Details