Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZCCHC13
Entrez Gene ID
101112704
Gene Name
zinc finger CCHC-type containing 13
Gene Aliases
-
Chromosome Location
Chr.X: 75811058 - 75811623 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004022196.4 XP_004022245.1 1 - 1 322 108

View Full Product Details