Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARL14
Entrez Gene ID
101105298
Gene Name
ADP ribosylation factor like GTPase 14
Gene Aliases
-
Chromosome Location
Chr.1: 247478837 - 247479857 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027966334.1 XP_027822135.1 1 - 1 372 114

View Full Product Details