Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HAO2
Entrez Gene ID
101110803
Gene Name
hydroxyacid oxidase 2
Gene Aliases
-
Chromosome Location
Chr.1: 101895564 - 101925651 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004002387.4 XP_004002436.2 7 - 8 1304 99
XM_012179575.3 XP_012034965.2 7 - 8 1281 99
XM_012179579.3 XP_012034969.2 8 - 9 1387 99

View Full Product Details