Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRIN3B
Entrez Gene ID
101112396
Gene Name
glutamate ionotropic receptor NMDA type subunit 3B
Gene Aliases
-
Chromosome Location
Chr.5: 45622020 - 45631258 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027970152.1 XP_027825953.1 3 - 4 2052 59

View Full Product Details