Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GAL3ST4
Entrez Gene ID
101106737
Gene Name
galactose-3-O-sulfotransferase 4
Gene Aliases
-
Chromosome Location
Chr.24: 36909030 - 36914762 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004021022.4 XP_004021071.2 3 - 4 722 103

View Full Product Details