Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANKK1
Entrez Gene ID
101109481
Gene Name
ankyrin repeat and kinase domain containing 1
Gene Aliases
-
Chromosome Location
Chr.15: 26473151 - 26487012 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027979116.1 XP_027834917.1 6 - 7 769 75

View Full Product Details