Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CEACAM16
Entrez Gene ID
101110784
Gene Name
CEA cell adhesion molecule 16, tectorial membrane component
Gene Aliases
-
Chromosome Location
Chr.14: 55975698 - 55982456 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027978452.1 XP_027834253.1 1 - 2 529 74

View Full Product Details