Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRGBP
Entrez Gene ID
101111383
Gene Name
MRG domain binding protein
Gene Aliases
-
Chromosome Location
Chr.13: 56935642 - 56940210 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027976970.1 XP_027832771.1 4 - 5 464 102

View Full Product Details