Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LACC1
Entrez Gene ID
101109625
Gene Name
laccase domain containing 1
Gene Aliases
-
Chromosome Location
Chr.10: 16258651 - 16271054 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012184387.2 XP_012039777.2 3 - 4 995 88
XM_027973535.1 XP_027829336.1 3 - 4 872 88

View Full Product Details