Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDCP2
Entrez Gene ID
101117722
Gene Name
CUB domain containing protein 2
Gene Aliases
-
Chromosome Location
Chr.1: 30259017 - 30280102 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012150012.3 XP_012005402.2 4 - 5 1120 106

View Full Product Details