Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OLFML3
Entrez Gene ID
101121985
Gene Name
olfactomedin like 3
Gene Aliases
-
Chromosome Location
Chr.1: 96121849 - 96124920 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004002351.4 XP_004002400.1 2 - 3 543 91

View Full Product Details