Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC15A5
Entrez Gene ID
101103710
Gene Name
solute carrier family 15 member 5
Gene Aliases
-
Chromosome Location
Chr.3: 213648523 - 213745649 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027965843.1 XP_027821644.1 3 - 4 759 92

View Full Product Details