Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHLDA1
Entrez Gene ID
101102304
Gene Name
pleckstrin homology like domain family A member 1
Gene Aliases
-
Chromosome Location
Chr.3: 119207892 - 119214079 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012173944.3 XP_012029334.3 1 - 2 1220 72

View Full Product Details