Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DIPK2B
Entrez Gene ID
101107931
Gene Name
divergent protein kinase domain 2B
Gene Aliases
CXHXorf36
Chromosome Location
Chr.X: 45748914 - 45789765 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004022021.4 XP_004022070.1 4 - 5 1147 52

View Full Product Details