Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TFF1
Entrez Gene ID
101104450
Gene Name
trefoil factor 1
Gene Aliases
-
Chromosome Location
Chr.1: 285943042 - 285947445 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004003363.3 XP_004003412.2 2 - 3 239 99

View Full Product Details