Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHRNA4
Entrez Gene ID
101118889
Gene Name
cholinergic receptor nicotinic alpha 4 subunit
Gene Aliases
-
Chromosome Location
Chr.13: 56544051 - 56556725 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027976432.1 XP_027832233.1 3 - 4 279 66

View Full Product Details