Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CXCL11
Entrez Gene ID
101108750
Gene Name
C-X-C motif chemokine ligand 11
Gene Aliases
-
Chromosome Location
Chr.6: 100777327 - 100779097 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015096572.2 XP_014952058.1 2 - 3 222 118
XM_027971095.1 XP_027826896.1 2 - 3 222 118

View Full Product Details