Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MSMB
Entrez Gene ID
101119365
Gene Name
microseminoprotein beta
Gene Aliases
-
Chromosome Location
Chr.25: 45939408 - 45949780 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004021566.4 XP_004021615.1 2 - 3 201 83

View Full Product Details